Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Chic2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK006962.1 | - | 4, 5 | 719 | ||
AK172289.1 | - | 4, 5 | 712 | |||
BC048435.1 | - | 5, 6 | 472 | |||
BC116971.1 | - | 4, 5 | 529 | 40126290 | ||
BC116973.1 | - | 4, 5 | 529 | 40126292 |