Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rnf145 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028862.3 | NP_083138.2 | 8, 9 | 1468 | ||
GenBank mRNA | AK014408.1 | - | 8, 9 | 1469 | ||
AK043002.1 | - | 8, 9 | 1455 | |||
BC040799.1 | - | 7, 8 | 1144 | 3988157 | ||
BC051064.1 | - | 1, 2 | 171 | |||
BC055485.1 | - | 2, 3 | 252 |