Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mamstr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172418.2 | NP_766006.2 | 7 | 813 | ||
GenBank mRNA | AK017382.1 | - | 6 | 1121 | ||
AK036109.1 | - | 4 | 931 | |||
AK052976.1 | - | 2 | 1442 | |||
BC107175.1 | - | 2 | 332 | |||
BC107176.1 | - | 2 | 334 | |||
BC119481.1 | - | 2 | 332 | |||
BC119482.1 | - | 2 | 332 | |||
BC127227.1 | - | 2 | 334 | |||
BC127228.1 | - | 2 | 334 | |||
DQ534901.1 | - | 7 | 797 |