Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kcnk16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029006.1 | NP_083282.1 | 5 | 762 | ||
GenBank mRNA | AK014626.1 | - | 631 |