Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pik3cb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029094.3 | NP_083370.2 | 6, 7 | 1121 | ||
GenBank mRNA | AK081350.1 | - | 6, 7 | 1316 | ||
AK090116.1 | - | 6, 7 | 998 | |||
AK154106.1 | - | 6, 7 | 1106 | |||
BC039650.1 | - | 6, 7 | 1084 | |||
CV558291.1 | - | 5, 6 | 871 |