Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sf3a3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029157.3 | NP_083433.1 | 3 | 203 | ||
GenBank mRNA | AK015776.1 | - | 3 | 182 | ||
AK030743.1 | - | 3 | 238 | |||
AK150733.1 | - | 3 | 201 | |||
AK151141.1 | - | 3 | 201 | |||
AK151382.1 | - | 3 | 201 | |||
AK161646.1 | - | 3 | 224 | |||
AK168966.1 | - | 3 | 227 | |||
AK169324.1 | - | 3 | 247 | |||
BC009141.1 | - | 3 | 203 | 3598647 | ||
BC092058.1 | - | 3 | 235 | 4017388 |