Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Taf1d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027261.3 | NP_081537.1 | 4, 5 | 730 | ||
NM_029248.2 | NP_083524.2 | 4, 5 | 730 | |||
GenBank mRNA | AK012656.1 | - | 2, 3 | 580 | ||
AK012662.1 | - | 4, 5 | 682 | |||
AK014920.1 | - | 4, 5 | 682 | |||
AK016109.1 | - | 4, 5 | 684 | |||
AK020036.1 | - | 3, 4 | 642 | |||
AK050165.1 | - | 4, 5 | 686 | |||
AK161656.1 | - | 4, 5 | 685 | |||
AK166329.1 | - | 4, 5 | 685 | |||
AK166633.1 | - | 4, 5 | 685 | |||
BC027813.1 | - | 2, 3 | 518 | |||
BC056964.1 | - | 3, 4 | 668 | |||
BC110660.1 | - | 3, 4 | 672 | 3158156 | ||
BC117068.1 | - | 3, 4 | 662 | 40126387 | ||
BC117070.1 | - | 3, 4 | 662 | 40126389 |