Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccdc83 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029256.3 | NP_083532.1 | 4 | 335 | ||
NM_183293.1 | NP_899116.1 | 4 | 335 | |||
GenBank mRNA | AK016080.1 | - | 4 | 337 | ||
AK016114.1 | - | 4 | 336 | |||
AK016535.1 | - | 4 | 322 | |||
BC120721.1 | - | 4 | 309 | 40129644 | ||
BC120723.1 | - | 4 | 309 | 40129646 | ||
BC145034.1 | - | 4 | 309 |