Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zbtb4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029348.2 | NP_083624.2 | 3 | 781 | ||
GenBank mRNA | AK009517.1 | - | 3 | 702 | ||
AK147212.1 | - | 3 | 735 | |||
AK163593.1 | - | 3 | 844 | |||
AK173196.1 | - | 1 | 535 | |||
BC112421.1 | - | 2 | 577 | |||
BC113187.1 | - | 2 | 577 |