Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ndufaf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127346.1 | NP_001120818.1 | 1 | 155 | ||
GenBank mRNA | AB183434.1 | - | 1 | 99 | ||
AK007894.1 | - | 1 | 146 | |||
AY262012.1 | - | 1 | 128 | |||
BC048512.1 | - | 1 | 114 | |||
BC116975.1 | - | 1 | 121 | 40126294 | ||
BC116977.1 | - | 1 | 121 | 40126296 | ||
BC144716.1 | - | 1 | 121 | |||
BY251841.1 | - | 1 | 156 |
Gene Symbol: B230220B15Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK080810.1 | - | 1 | 157 |