Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Depdc1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001172092.1 | NP_001165563.1 | 6, 7 | 858 | ||
NM_001172093.1 | NP_001165564.1 | 6, 7 | 858 | |||
NM_029523.3 | NP_083799.2 | 6, 7 | 858 | |||
GenBank mRNA | AK050456.1 | - | 6, 7 | 795 | ||
AK088719.1 | - | 6, 7 | 834 | |||
AK146458.1 | - | 6, 7 | 827 | |||
BC023934.1 | - | 6, 7 | 851 | 5342167 |