Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rab2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172601.3 | NP_766189.1 | 7 | 671 | ||
GenBank mRNA | AB232586.1 | - | 7 | 517 | ||
AK005230.1 | - | 7 | 574 | |||
AK038393.1 | - | 7 | 606 | |||
AK044202.1 | - | 7 | 548 | |||
AK160249.1 | - | 7 | 822 | |||
AK160351.1 | - | 7 | 537 | |||
BC033312.1 | - | 7 | 568 | 4481516 | ||
BC046334.1 | - | 7 | 613 | 6492838 | ||
BC054719.1 | - | 7 | 604 | |||
BC061513.1 | - | 3 | 620 |