Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Snip1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175246.4 | NP_780455.2 | 3 | 604 | ||
GenBank mRNA | AK010794.1 | - | 3 | 605 | ||
AK040571.1 | - | 3 | 605 | |||
AK049254.1 | - | 3 | 605 | |||
AK049318.1 | - | 3 | 605 | |||
AK078163.1 | - | 3 | 682 | |||
AK132768.1 | - | 3 | 582 | |||
AK143801.1 | - | 3 | 604 | |||
BC064067.1 | - | 3 | 577 | 949686 |