Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ssbp4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133772.2 | NP_598533.2 | 3, 4 | 329 | ||
GenBank mRNA | AK004835.1 | - | 3, 4 | 441 | ||
AK154336.1 | - | 3, 4 | 330 | |||
AK157387.1 | - | 3, 4 | 318 | |||
BC002150.1 | - | 3, 4 | 288 | 3484896 | ||
CX214457.1 | - | 3, 4 | 329 | |||
CX237617.1 | - | 2, 3 | 111 |