Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ticrr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029835.1 | NP_084111.1 | 4 | 1398 | ||
GenBank mRNA | AK041726.1 | - | 4 | 1348 | ||
AK051639.1 | - | 4 | 1361 | |||
AK054492.1 | - | 4 | 1348 | |||
AK137597.1 | - | 4 | 1364 | |||
AK157802.1 | - | 3 | 2901 | |||
BC158103.1 | - | 4 | 1362 | 9088180 |