Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mmab | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029956.3 | NP_084232.1 | 5 | 426 | ||
GenBank mRNA | AK020286.1 | - | 5 | 426 | ||
AK034288.1 | - | 5 | 391 | |||
AK088785.1 | - | 5 | 418 | |||
AK142286.1 | - | 5 | 418 | |||
AK148421.1 | - | 5 | 461 | |||
BC022159.1 | - | 5 | 380 | |||
BC057558.1 | - | 5 | 426 | 6415979 |