Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sfi1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030207.2 | NP_084483.2 | 2, 3 | 211 | ||
GenBank mRNA | AK046135.1 | - | 2, 3 | 208 | ||
AK046373.1 | - | 2, 3 | 208 | |||
AK082866.1 | - | 2, 3 | 173 | |||
AK129159.1 | - | 2, 3 | 178 | |||
AK133558.1 | - | 2, 3 | 159 | |||
AK170090.1 | - | 2, 3 | 171 | |||
BC046305.1 | - | 2, 3 | 159 | 6493676 | ||
BC099677.1 | - | 2, 3 | 187 | |||
BY303229.1 | - | 2, 3 | 212 |