Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sfi1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030207.2 | NP_084483.2 | 22 | 2309 | ||
GenBank mRNA | AK019095.1 | - | 2 | 98 | ||
AK046135.1 | - | 16 | 1719 | |||
AK046373.1 | - | 16 | 1719 | |||
AK133558.1 | - | 22 | 2257 | |||
AK170090.1 | - | 17 | 1749 | |||
BC026390.1 | - | 11 | 950 | |||
BC046305.1 | - | 15 | 1477 | 6493676 | ||
BC048950.1 | - | 5 | 337 | |||
BC099677.1 | - | 20 | 2059 |