Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc26a5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289787.1 | NP_001276716.1 | 2 | 228 | ||
NM_001289788.1 | NP_001276717.1 | 2 | 228 | |||
NM_030727.5 | NP_109652.3 | 3 | 303 | |||
GenBank mRNA | AF529192.1 | - | 3 | 303 | ||
AY024359.2 | - | 1 | 69 | |||
BC108986.1 | - | 2 | 220 | 40047724 | ||
BC108987.1 | - | 2 | 220 | 40047725 |