Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Csnk1a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146087.2 | NP_666199.1 | 8 | 936 | ||
GenBank mRNA | AK076070.1 | - | 8 | 1203 | ||
AK076264.1 | - | 8 | 1195 | |||
AK078398.1 | - | 8 | 1185 | |||
AK141421.1 | - | 8 | 1191 | |||
AK146873.1 | - | 8 | 1206 | |||
AK147165.1 | - | 8 | 1181 | |||
AK152387.1 | - | 8 | 1181 | |||
AK152793.1 | - | 8 | 1201 | |||
AK159939.1 | - | 5 | 405 | |||
AK166318.1 | - | 8 | 1196 | |||
AK166429.1 | - | 8 | 1178 | |||
AK167550.1 | - | 8 | 1204 | |||
AK180731.1 | - | 5 | 379 | |||
AK183976.1 | - | 5 | 371 | |||
AK219793.1 | - | 5 | 347 | |||
BC002171.1 | - | 2 | 84 | |||
BC019740.1 | - | 8 | 936 | 3487913 | ||
BC025439.1 | - | 8 | 883 | 5038433 | ||
BC048081.1 | - | 8 | 969 | 6309504 |