Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aff4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033565.2 | NP_291043.1 | 11 | 2243 | ||
GenBank mRNA | AF190449.1 | - | 11 | 2185 | ||
AK033163.1 | - | 1 | 65 | |||
AK044058.1 | - | 11 | 2128 | |||
AK054401.1 | - | 11 | 2093 | |||
AK163213.1 | - | 1 | 701 | |||
BC138999.1 | - | 11 | 1916 | 8862021 | ||
BC145490.1 | - | 10 | 1829 | 9054053 |