Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nipa2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256130.1 | NP_001243059.1 | 5, 6 | 788 | ||
NM_001256131.1 | NP_001243060.1 | 4, 5 | 667 | |||
NM_001256132.1 | NP_001243061.1 | 5, 6 | 592 | |||
NM_001256133.1 | NP_001243062.1 | 3, 4 | 242 | |||
NM_023647.6 | NP_076136.2 | 5, 6 | 1542 | |||
GenBank mRNA | AB041581.1 | - | 5, 6 | 725 | ||
AK008106.1 | - | 5, 6 | 775 | |||
AK011248.1 | - | 4, 5 | 612 | |||
AK012859.1 | - | 5, 6 | 726 | |||
AK028734.1 | - | 5, 6 | 1500 | |||
AK041427.1 | - | 5, 6 | 789 | |||
AK076051.1 | - | 5, 6 | 745 | |||
AK076184.1 | - | 5, 6 | 729 | |||
AK154755.1 | - | 5, 6 | 746 | |||
AV495155.1 | - | 3, 4 | 185 | |||
BC038499.1 | - | 5, 6 | 730 | 4482150 |