Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trim11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290988.1 | NP_001277917.1 | 7 | 1323 | ||
NM_053168.2 | NP_444398.1 | 6 | 1275 | |||
GenBank mRNA | AF220124.1 | - | 6 | 1252 | ||
AK051266.1 | - | 5 | 2492 | |||
AK139477.1 | - | 6 | 1260 | |||
AK157332.1 | - | 6 | 1268 | |||
AK170656.1 | - | 6 | 1241 | |||
AK170855.1 | - | 4 | 2604 | |||
BC020102.1 | - | 7 | 1301 | 4014766 |