Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Srd5a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053188.2 | NP_444418.1 | 3, 4 | 634 | ||
GenBank mRNA | AB049456.1 | - | 3, 4 | 634 | ||
AK138946.1 | - | 3, 4 | 575 | |||
BC125510.1 | - | 3, 4 | 618 | 40130025 |