Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmem40 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001168256.1 | NP_001161728.1 | 3 | 292 | ||
NM_001168257.1 | NP_001161729.1 | 3 | 292 | |||
NM_144805.2 | NP_659054.2 | 3 | 160 | |||
GenBank mRNA | BC019416.1 | - | 3 | 122 | 5151159 | |
BY104106.1 | - | 3 | 147 | |||
BY279100.1 | - | 3 | 292 | |||
CT010303.1 | - | 2 | 90 | |||
CV561368.1 | - | 3 | 234 | |||
CX564997.1 | - | 3 | 191 |