Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rab3gap2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163754.1 | NP_001157226.1 | 35 | 4136 | ||
GenBank mRNA | AK030955.1 | - | 24 | 3046 | ||
AK031191.1 | - | 25 | 3045 | |||
AK037092.1 | - | 3 | 450 | |||
AK048759.1 | - | 11 | 2477 | |||
AK122385.1 | - | 20 | 4824 | |||
AK149265.1 | - | 12 | 1307 | |||
BC035944.1 | - | 26 | 3126 | |||
BC052505.1 | - | 10 | 1234 | |||
BC057872.1 | - | 26 | 3154 | 5294131 |