Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ankrd50 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK147106.1 | - | 1 | 1251 | ||
AK162481.1 | - | 1 | 959 | |||
AK194127.1 | - | 1 | 145 | |||
AK220375.1 | - | 1 | 1248 | |||
BC060298.1 | - | 1 | 43 | |||
BC145902.1 | - | 1 | 930 | 40131205 |