Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Plod1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053827.2 | NP_446279.2 | 5, 6 | 629 | ||
GenBank mRNA | CA513298.1 | - | 3, 4 | 348 | ||
FQ074486.1 | - | 5, 6 | 628 | |||
FQ202794.1 | - | 5, 6 | 611 |