Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slco6b1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133412.1 | NP_596903.1 | 3, 4 | 1010 | ||
XM_006245576.2 | XP_006245638.1 | 3, 4 | 1080 | |||
GenBank mRNA | AF321415.1 | - | 3, 4 | 1010 |
Gene Symbol: LOC102554121 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006245586.2 | XP_006245648.1 | 3, 4 | 1081 |