Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trak2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133560.2 | NP_598244.2 | 3 | 525 | ||
XM_006244954.2 | XP_006245016.1 | 3 | 602 | |||
XM_008767072.1 | XP_008765294.1 | 3 | 603 | |||
XM_008767073.1 | XP_008765295.1 | 3 | 780 | |||
GenBank mRNA | AF474163.1 | - | 2 | 155 | ||
AJ288898.2 | - | 3 | 525 | |||
BC088393.1 | - | 3 | 524 | 7124967 |