Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Asgr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012503.2 | NP_036635.1 | 5 | 378 | ||
XM_006246579.1 | XP_006246641.1 | 5 | 490 | |||
XM_008767768.1 | XP_008765990.1 | 5 | 510 | |||
XM_008767769.1 | XP_008765991.1 | 5 | 454 | |||
GenBank mRNA | BC088154.1 | - | 5 | 378 | 7372227 | |
FQ209763.1 | - | 5 | 414 | |||
FQ218553.1 | - | 5 | 454 | |||
FQ218644.1 | - | 5 | 414 | |||
FQ218824.1 | - | 5 | 414 | |||
FQ219107.1 | - | 5 | 451 | |||
FQ219710.1 | - | 5 | 495 | |||
M21770.1 | - | 4 | 280 |