Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fgfr2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109892.1 | NP_001103362.1 | 7 | 1544 | ||
NM_001109893.1 | NP_001103363.1 | 6 | 1277 | |||
NM_001109894.1 | NP_001103364.1 | 6 | 1277 | |||
NM_001109895.1 | NP_001103365.1 | 5 | 1199 | |||
NM_001109896.1 | NP_001103366.1 | 5 | 1199 | |||
NM_012712.1 | NP_036844.1 | 7 | 1544 | |||
XM_006230389.2 | XP_006230451.1 | 7 | 1545 | |||
XM_006230390.2 | XP_006230452.1 | 7 | 1545 | |||
XM_006230392.2 | XP_006230454.1 | 7 | 1545 | |||
XM_006230393.2 | XP_006230455.1 | 5 | 1200 | |||
XM_008759861.1 | XP_008758083.1 | 6 | 3701 | |||
GenBank mRNA | AF294649.1 | - | 3 | 382 | ||
AF294650.1 | - | 3 | 382 | |||
L19104.1 | - | 6 | 980 | |||
L19105.1 | - | 6 | 982 | |||
L19107.1 | - | 5 | 713 | |||
L19108.1 | - | 4 | 635 | |||
L19111.1 | - | 4 | 635 | |||
L19112.1 | - | 5 | 705 | |||
Z35138.1 | - | 4 | 553 |