Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gata1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012764.1 | NP_036896.1 | 397 | |||
GenBank mRNA | D13518.1 | - | 397 |
Gene Symbol: NEWGENE_2663 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008773093.1 | XP_008771315.1 | 3 | 418 |