Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Map2k4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001030023.1 | NP_001025194.1 | 8 | 906 | ||
GenBank mRNA | AY880882.1 | - | 8 | 906 | ||
KC795688.1 | - | 9 | 939 |