Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ttll6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008768194.1 | XP_008766416.1 | 6 | 788 | ||
XM_008768195.1 | XP_008766417.1 | 6 | 788 |