Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Qtrtd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006248320.2 | XP_006248382.1 | 5 | 583 | ||
XM_008768759.1 | XP_008766981.1 | 5 | 583 | |||
XM_008768760.1 | XP_008766982.1 | 5 | 583 | |||
XM_008768761.1 | XP_008766983.1 | 5 | 583 | |||
XM_008768762.1 | XP_008766984.1 | 3 | 201 |