Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Taf6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001044225.1 | NP_001037690.1 | 12, 13 | 1368 | ||
XM_008769061.1 | XP_008767283.1 | 12, 13 | 1408 | |||
XM_008769062.1 | XP_008767284.1 | 12, 13 | 1407 | |||
GenBank mRNA | BC100108.1 | - | 12, 13 | 1368 | 7379862 | |
D49446.1 | - | 11, 12 | 1339 |