Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Camsap2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134503.1 | NP_001127975.1 | 12 | 3259 | ||
XM_008769549.1 | XP_008767771.1 | 12 | 2754 | |||
XM_008769550.1 | XP_008767772.1 | 11 | 2721 | |||
XM_008769551.1 | XP_008767773.1 | 11 | 2721 |