Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gnl3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175580.2 | NP_783170.1 | 7 | 620 | ||
GenBank mRNA | AY181024.1 | - | 7 | 620 | ||
BC072500.1 | - | 5 | 1143 | |||
BC093602.1 | - | 7 | 674 | 7316170 |