Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Agpat6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001047849.1 | NP_001041314.1 | 5, 6 | 1526 | ||
GenBank mRNA | BC082092.1 | - | 2, 3 | 372 | ||
BC161809.1 | - | 4, 5 | 812 | 8374387 | ||
DQ480750.1 | - | 5, 6 | 1526 |