Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam65a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100660.1 | NP_001094130.1 | 15, 16 | 2855 | ||
XM_006255470.2 | XP_006255532.1 | 15, 16 | 3204 | |||
XM_006255471.2 | XP_006255533.1 | 15, 16 | 3316 | |||
XM_006255472.2 | XP_006255534.1 | 15, 16 | 3305 | |||
XM_006255473.2 | XP_006255535.1 | 15, 16 | 3202 | |||
XM_006255474.2 | XP_006255536.1 | 15, 16 | 3204 | |||
XM_006255475.2 | XP_006255537.1 | 15, 16 | 2868 | |||
GenBank mRNA | AY724537.1 | - | 7, 8 | 2084 | ||
BC099098.1 | - | 5, 6 | 1822 |