Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Akap13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106271.2 | NP_001099741.2 | 7 | 1739 | ||
XM_006229382.2 | XP_006229444.1 | 7 | 2550 | |||
XM_006229384.2 | XP_006229446.1 | 7 | 2550 | |||
XM_006229386.2 | XP_006229448.1 | 7 | 2550 |