Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tas2r134 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013915.1 | NP_001013937.1 | 1 | 883 | ||
GenBank mRNA | AY916511.1 | - | 1 | 802 |