Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pcolce | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019237.1 | NP_062110.1 | 4 | 628 | ||
GenBank mRNA | AB008534.1 | - | 4 | 615 | ||
AF016503.1 | - | 4 | 622 | |||
BC086572.1 | - | 5 | 684 | 7304047 | ||
FQ209381.1 | - | 4 | 664 | |||
U94710.1 | - | 4 | 628 |