siRNA Details
Gene Symbol: Nup214 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001168559.1 | NP_001162031.1 | 26 | 5984 | ||
GenBank mRNA | FQ226859.1 | - | 5 | 697 |

Gene Symbol: Nup214 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001168559.1 | NP_001162031.1 | 26 | 5984 | ||
GenBank mRNA | FQ226859.1 | - | 5 | 697 |