Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lmod2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100964.1 | NP_001094434.1 | 2 | 1272 | ||
GenBank mRNA | AB331240.1 | - | 2 | 1148 | ||
BC128755.1 | - | 2 | 1272 | 7104774 |
Gene Symbol: LOC100909784 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003749707.3 | XP_003749755.1 | 2 | 1344 |