Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Grik3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001112716.1 | NP_001106187.1 | 6 | 974 | ||
NM_181373.3 | NP_852038.2 | 6 | 974 | |||
XM_006238857.2 | XP_006238919.1 | 6 | 1565 | |||
GenBank mRNA | AF027331.1 | - | 6 | 977 | ||
M83552.1 | - | 6 | 750 | |||
Z11716.1 | - | 6 | 977 |