Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cyp4f39 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006241090.1 | XP_006241152.1 | 4 | 449 | ||
XM_006241091.2 | XP_006241153.1 | 4 | 558 | |||
XM_006241092.1 | XP_006241154.1 | 3 | 361 | |||
XM_008765183.1 | XP_008763405.1 | 3 | 789 | |||
XM_234837.7 | XP_234837.3 | 4 | 890 | |||
GenBank mRNA | FQ210490.1 | - | 4 | 454 |