Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam46a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106844.1 | NP_001100314.1 | 2 | 787 | ||
XM_006243474.2 | XP_006243536.1 | 2 | 770 |