Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atrip | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106859.1 | NP_001100329.1 | 7, 8 | 1167 | ||
XM_006243764.2 | XP_006243826.1 | 7, 8 | 1169 | |||
XM_006243766.2 | XP_006243828.1 | 7, 8 | 1168 | |||
XM_006243767.2 | XP_006243829.1 | 7, 8 | 1168 |
Gene Symbol: LOC100911807 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003750589.3 | XP_003750637.1 | 7, 8 | 2162 |